missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-2-naphthol, 98%

Catalog No. AC177470500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC177470500 50 g Glass bottle
AC177470100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177470500 Supplier Thermo Scientific Chemicals Supplier No. 177470500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1125-78-6 | C10H12O | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1125-78-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.2
MDL Number MFCD00001738
InChI Key UMKXSOXZAXIOPJ-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
PubChem CID 14305
ChEBI CHEBI:34448
IUPAC Name 5,6,7,8-tetrahydronaphthalen-2-ol
SMILES C1CCC2=C(C1)C=CC(=C2)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58°C to 62°C
Color Beige
Boiling Point 275°C to 276°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Beilstein 06, 579
Formula Weight 148.2
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 5,6,7,8-Tetrahydro-2-naphthol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-410-4
RTECSNumber QL5300000

RUO – Research Use Only