missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-2-methylquinoline 98.0+%, TCI America™

Catalog No. T140810ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T140810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T140810ML Supplier TCI America Supplier No. T140810ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity building block for laboratory research, with >98.0% purity as determined by GC and nonaqueous titration.

Chemical Identifiers

CAS 2617-98-3
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00191581
InChI Key AUSMPOZTRKHFPG-UHFFFAOYSA-N
Synonym 5,6,7,8-Tetrahydroquinaldine
PubChem CID 12229420
IUPAC Name 2-methyl-5,6,7,8-tetrahydroquinoline
SMILES CC1=NC2=C(CCCC2)C=C1

Specifications

Color Colorless
Boiling Point 225°C
Quantity 10 mL
Formula Weight 147.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydro-2-methylquinoline