accessibility menu, dialog, popup

UserName

CAS RN 2617-98-3

IUPAC Name: 2-methyl-5,6,7,8-tetrahydroquinoline
Synonyms: 2-methyl-5,6,7,8-tetrahydroquinoline
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: AUSMPOZTRKHFPG-UHFFFAOYSA-N
SMILES: CC1=NC2=C(CCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results