missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-1-naphthylamine 98.0+%, TCI America™

Catalog No. p-7117175
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09955ML 5 mL
A099525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09955ML Supplier TCI America Supplier No. A09955ML

Chemical Identifiers

CAS 2217-41-6
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00001736
InChI Key SODWJACROGQSMM-UHFFFAOYSA-N
Synonym 5,6,7,8-tetrahydro-1-naphthylamine, 5-aminotetralin, 1-amino-5,6,7,8-tetrahydronaphthalene, 5-tetralylamine, 1-naphthalenamine, 5,6,7,8-tetrahydro, 1-naphthylamine, 5,6,7,8-tetrahydro, 5-amino-1,2,3,4-tetrahydronaphthalene, tetralin-5-amine, alpha-tetrahydronaphthylamine, 5,6,7,8-tetrahydro-1-naphthalenamine
PubChem CID 16671
IUPAC Name 5,6,7,8-tetrahydronaphthalen-1-amine
SMILES NC1=CC=CC2=C1CCCC2

Specifications

Boiling Point 275°C
Quantity 5 mL
Formula Weight 147.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydro-1-naphthylamine
RTECSNumber QM4780000
TSCA No