missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6,7,8-Tetrahydro-1-naphthol 98.0+%, TCI America™
Supplier: TCI America T281625G
Specifications
| 5,6,7,8-Tetrahydro-1-naphthol | |
| 70°C | |
| C10H12O | |
| 25 g | |
| SCWNNOCLLOHZIG-UHFFFAOYSA-N | |
| 5,6,7,8-tetrahydronaphthalen-1-ol | |
| 68258 | |
| 148.21 | |
| Crystalline Powder |
| 529-35-1 | |
| 265°C | |
| MFCD00001734 | |
| 5,6,7,8-tetrahydro-1-naphthol, 5-hydroxytetralin, 1-naphthalenol, 5,6,7,8-tetrahydro, tetralin-5-ol, 5,6,7,8-tetrahydro-alpha-naphthol, unii-1r7b5i98hv, 5,6,7,8-tetrahydronaphthol, 5,6,7,8-tetrahydro-1-naphthalenol, 5,6,7,8-tetrahydro-naphthalen-1-ol, tetrahydro-.alpha.-naphthol | |
| OC1=CC=CC2=C1CCCC2 | |
| 148.21 | |
| CHEBI:45900 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 529-35-1 | |
| 148.21 | |
| SCWNNOCLLOHZIG-UHFFFAOYSA-N | |
| 68258 | |
| 5,6,7,8-tetrahydronaphthalen-1-ol |
| C10H12O | |
| MFCD00001734 | |
| 5,6,7,8-tetrahydro-1-naphthol, 5-hydroxytetralin, 1-naphthalenol, 5,6,7,8-tetrahydro, tetralin-5-ol, 5,6,7,8-tetrahydro-alpha-naphthol, unii-1r7b5i98hv, 5,6,7,8-tetrahydronaphthol, 5,6,7,8-tetrahydro-1-naphthalenol, 5,6,7,8-tetrahydro-naphthalen-1-ol, tetrahydro-.alpha.-naphthol | |
| CHEBI:45900 | |
| OC1=CC=CC2=C1CCCC2 |
Safety and Handling
TSCA : Yes