missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one Hydrochloride 97.0+%, TCI America™

Catalog No. T29841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29841G 1 g
T29845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29841G Supplier TCI America Supplier No. T29841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115473-15-9
Molecular Formula C7H10ClNOS
Molecular Weight (g/mol) 191.673
MDL Number MFCD11111130
InChI Key PUQKTVAKLPDUAW-UHFFFAOYSA-N
PubChem CID 23435874
IUPAC Name 5,6,7,7a-tetrahydro-4H-thieno[3,2-c]pyridin-2-one;hydrochloride
SMILES C1CNCC2=CC(=O)SC21.Cl

Specifications

Melting Point 210°C
Quantity 1 g
Formula Weight 191.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6,7,7a-Tetrahydrothieno[3,2-c]pyridin-2(4H)-one Hydrochloride
TSCA No
Recommended Storage Freezer