missing translation for 'onlineSavingsMsg'
Learn More

5,6,11,12-Tetraphenylnaphthacene 98.0+%, TCI America™

Catalog No. p-7136345
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0561100MG 100 mg
T05611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0561100MG Supplier TCI America Supplier No. T0561100MG
Only null left

Chemical Identifiers

CAS 517-51-1
Molecular Formula C42H28
Molecular Weight (g/mol) 532.686
MDL Number MFCD00003703
InChI Key YYMBJDOZVAITBP-UHFFFAOYSA-N
Synonym rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl
PubChem CID 68203
IUPAC Name 5,6,11,12-tetraphenyltetracene
SMILES C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8

Specifications

Melting Point 331°C
Quantity 100 mg
Formula Weight 532.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6,11,12-Tetraphenylnaphthacene