missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6,11,12-Tetraphenylnaphthacene (purified by sublimation) 99.0+%, TCI America™
Supplier: TCI America T22331G
Specifications
| 5,6,11,12-Tetraphenylnaphthacene (purified by sublimation) | |
| 331°C | |
| MFCD00003703 | |
| rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl | |
| C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8 | |
| 532.686 | |
| 532.69 | |
| Crystalline Powder |
| 517-51-1 | |
| C42H28 | |
| 1 g | |
| YYMBJDOZVAITBP-UHFFFAOYSA-N | |
| 5,6,11,12-tetraphenyltetracene | |
| 68203 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 517-51-1 | |
| 532.686 | |
| YYMBJDOZVAITBP-UHFFFAOYSA-N | |
| 68203 | |
| C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=C(C5=CC=CC=C5C(=C24)C6=CC=CC=C6)C7=CC=CC=C7)C8=CC=CC=C8 |
| C42H28 | |
| MFCD00003703 | |
| rubrene, 5,6,11,12-tetraphenylnaphthacene, rubren, naphthacene, 5,6,11,12-tetraphenyl, rubrene, powder, acmc-209kvj, 5,11,12-tetraphenyltetracene, 5,11,12-tetraphenylnaphthacene, naphthacene,6,11,12-tetraphenyl | |
| 5,6,11,12-tetraphenyltetracene |
Safety and Handling
TSCA : Yes