missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dithiobis(2-nitrobenzoic acid), 99%

Catalog No. AAA1433103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1433103 1 g
AAA1433106 5 g
AAA1433114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1433103 Supplier Thermo Scientific Chemicals Supplier No. A1433103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69-78-3 | C14H8N2O8S2 | 396.34 g/mol

Reagent for determination of sufhydryl groups.5,5'-dithiobis-(2-nitrobenzoic acid) is widely utilized as a non-fluorescent probe to quantify the number or the concentration of thiol groups in a sample. It acts as a cleavable cross-linking agent. It finds an important application as a sensitive reagent for determining free sulfhydryl content in proteins, peptides, and tissues. It is also employed in high-performance liquid chromatography (HPLC) to determine the alkylthiol content.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for determination of sufhydryl groups.5,5′-dithiobis-(2-nitrobenzoic acid) is widely utilized as a non-fluorescent probe to quantify the number or the concentration of thiol groups in a sample. It acts as a cleavable cross-linking agent. It finds an important application as a sensitive reagent for determining free sulfhydryl content in proteins, peptides, and tissues. It is also employed in high-performance liquid chromatography (HPLC) to determine the alkylthiol content.

Solubility
Soluble in water and ethanol.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 69-78-3
Molecular Formula C14H8N2O8S2
Molecular Weight (g/mol) 396.34
MDL Number MFCD00007140
InChI Key KIUMMUBSPKGMOY-UHFFFAOYSA-N
Synonym dtnb, 5,5'-dithiobis 2-nitrobenzoic acid, ellman's reagent, 3-carboxy-4-nitrophenyl disulfide, dithionitrobenzoic acid, dithiobisnitrobenzoic acid, 5,5'-disulfanediylbis 2-nitrobenzoic acid, benzoic acid, 3,3'-dithiobis 6-nitro, 3,3'-dithiobis 6-nitrobenzoic acid, 5,5'-dithio-bis 2-nitrobenzoic acid
PubChem CID 6254
ChEBI CHEBI:86228
IUPAC Name 5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
SMILES OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 1 g
Beilstein 1896821
Solubility Information Soluble in water and ethanol.
Formula Weight 396.36
Percent Purity 99%
Chemical Name or Material 5,5'-Dithiobis(2-nitrobenzoic acid)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-714-4
RTECSNumber DG9650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only