missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Dimethyl-2,4-oxazolidinedione 99.0+%, TCI America™
$171.49 - $648.70
Chemical Identifiers
| CAS | 695-53-4 |
|---|---|
| Molecular Formula | C5H7NO3 |
| Molecular Weight (g/mol) | 129.12 |
| MDL Number | MFCD00005379 |
| InChI Key | JYJFNDQBESEHJQ-UHFFFAOYSA-N |
| Synonym | dimethadione, 5,5-dimethyloxazolidine-2,4-dione, 5,5-dimethyl-2,4-oxazolidinedione, propazone, dimethadion, eupractone, dimethyloxazolidinedione, 2,4-oxazolidinedione, 5,5-dimethyl, 5,5-dimethyloxazolidinedione, dimetadione dcit |
| PubChem CID | 3081 |
| IUPAC Name | 5,5-dimethyl-1,3-oxazolidine-2,4-dione |
| SMILES | CC1(C)OC(=O)NC1=O |
Chemical Identifiers
| 695-53-4 | |
| 129.12 | |
| JYJFNDQBESEHJQ-UHFFFAOYSA-N | |
| 3081 | |
| CC1(C)OC(=O)NC1=O |
| C5H7NO3 | |
| MFCD00005379 | |
| dimethadione, 5,5-dimethyloxazolidine-2,4-dione, 5,5-dimethyl-2,4-oxazolidinedione, propazone, dimethadion, eupractone, dimethyloxazolidinedione, 2,4-oxazolidinedione, 5,5-dimethyl, 5,5-dimethyloxazolidinedione, dimetadione dcit | |
| 5,5-dimethyl-1,3-oxazolidine-2,4-dione |
Specifications
| 695-53-4 | |
| White-Yellow | |
| C5H7NO3 | |
| 5 g | |
| JYJFNDQBESEHJQ-UHFFFAOYSA-N | |
| 5,5-dimethyl-1,3-oxazolidine-2,4-dione | |
| 3081 | |
| ≥99.0% (GC,T) | |
| 5,5-Dimethyl-2,4-oxazolidinedione |
| 79°C | |
| 137°C | |
| MFCD00005379 | |
| dimethadione, 5,5-dimethyloxazolidine-2,4-dione, 5,5-dimethyl-2,4-oxazolidinedione, propazone, dimethadion, eupractone, dimethyloxazolidinedione, 2,4-oxazolidinedione, 5,5-dimethyl, 5,5-dimethyloxazolidinedione, dimetadione dcit | |
| CC1(C)OC(=O)NC1=O | |
| 129.12 | |
| 129.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0180
RTECSNumber : RP9100000
TSCA : Yes