CAS RN 695-53-4
CAS RN 695-53-4
IUPAC Name:
5,5-dimethyl-1,3-oxazolidine-2,4-dione
Synonyms:
5,5-dimethyl-1,3-oxazolidine-2,4-dione
Molecular Weight (g/mol):
129.12
Molecular Formula:
C5H7NO3
InChi Key:
JYJFNDQBESEHJQ-UHFFFAOYSA-N
SMILES:
CC1(C)OC(=O)NC1=O
1
–
1
of
1
results
1
5,5-Dimethyl-2,4-oxazolidinedione 99.0+%, TCI America™
CAS: 695-53-4 Molecular Formula: C5H7NO3 Molecular Weight (g/mol): 129.12 MDL Number: MFCD00005379 InChI Key: JYJFNDQBESEHJQ-UHFFFAOYSA-N Synonym: dimethadione,5,5-dimethyloxazolidine-2,4-dione,5,5-dimethyl-2,4-oxazolidinedione,propazone,dimethadion,eupractone,dimethyloxazolidinedione,2,4-oxazolidinedione, 5,5-dimethyl,5,5-dimethyloxazolidinedione,dimetadione dcit PubChem CID: 3081 IUPAC Name: 5,5-dimethyl-1,3-oxazolidine-2,4-dione SMILES: CC1(C)OC(=O)NC1=O