missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 27°C |
| Color | White-Yellow |
| Boiling Point | 110°C |
| Quantity | 5 g |
| Formula Weight | 263.92 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 5,5-Dimethoxy-1,2,3,4-tetrachlorocyclopentadiene |
Safety and Handling
| TSCA | No |