missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dibromomeldrum's Acid (=5,5-Dibromo-2,2-dimethyl-4,6-dioxy-1,3-dioxane) 98.0+%, TCI America™

Catalog No. D17105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17105G 5 g
D171025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17105G Supplier TCI America Supplier No. D17105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66131-14-4
Molecular Formula C6H6Br2O4
Molecular Weight (g/mol) 301.918
MDL Number MFCD00059808
InChI Key HPBNIRVIOCWRDC-UHFFFAOYSA-N
Synonym 5,5-Dibromo-2,2-dimethyl-4,6-dioxo-1,3-dioxane, Dibromomalonic Acid Cyclic Isopropylidine Ester, cycl-Isopropylidene Dibromomalonate
PubChem CID 11748639
IUPAC Name 5,5-dibromo-2,2-dimethyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)C(C(=O)O1)(Br)Br)C

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 5 g
Formula Weight 301.92
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Dibromomeldrum′s Acid (=5,5-Dibromo-2,2-dimethyl-4,6-dioxy-1,3-dioxane)
TSCA No
Recommended Storage Freezer