missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dibromo-3,3'-dihexyl-2,2'-bithiophene 97.0+%, TCI America™

Catalog No. D41835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41835G 5 g
D41831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41835G Supplier TCI America Supplier No. D41835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 170702-05-3
Molecular Formula C20H28Br2S2
Molecular Weight (g/mol) 492.372
InChI Key MMSTXAGVEKUBED-UHFFFAOYSA-N
PubChem CID 15185062
IUPAC Name 5-bromo-2-(5-bromo-3-hexylthiophen-2-yl)-3-hexylthiophene
SMILES CCCCCCC1=C(SC(=C1)Br)C2=C(C=C(S2)Br)CCCCCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 492.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5,5′-Dibromo-3,3′-dihexyl-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator