accessibility menu, dialog, popup

UserName

CAS RN 170702-05-3

IUPAC Name: 5,5'-dibromo-3,3'-dihexyl-2,2'-bithiophene
Synonyms: 5,5'-dibromo-3,3'-dihexyl-2,2'-bithiophene
Molecular Weight (g/mol): 492.37
Molecular Formula: C20H28Br2S2
InChi Key: MMSTXAGVEKUBED-UHFFFAOYSA-N
SMILES: CCCCCCC1=C(SC(Br)=C1)C1=C(CCCCCC)C=C(Br)S1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results