missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Bromo-2-thienyl)isoxazole, ≥97%, Thermo Scientific™

Catalog No. p-3110770
Change view
Click to view available options
Quantity:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC16010DA 1g
CC16010EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC16010DA Supplier Thermo Scientific Chemicals Supplier No. CC16010DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138716-31-1
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.079
MDL Number MFCD03618406
InChI Key FCUZGSQWFAXBSB-UHFFFAOYSA-N
Synonym 2-bromo-5-isoxazol-5-yl thiophene, 5-5-bromo-2-thienyl isoxazole, 5-5-bromothiophen-2-yl-1,2-oxazole, 5-bromo-2-isoxazol-5-ylthiophene, 5-5-bromothien-2-yl isoxazole, 5-5-bromo-2-thiophenyl isoxazole, 5-5-bromothiophen-2-yl isoxazole, 5-5-bromothien-2-yl-1,2-oxazole, 5-5-bromanylthiophen-2-yl-1,2-oxazole
PubChem CID 2757026
IUPAC Name 5-(5-bromothiophen-2-yl)-1,2-oxazole
SMILES C1=C(SC(=C1)Br)C2=CC=NO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 230.08
Percent Purity 97%
Chemical Name or Material 5-(5-Bromo-2-thienyl)isoxazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.