accessibility menu, dialog, popup

UserName

SBrNO

CAS RN 138716-31-1

IUPAC Name: 5-(5-bromothiophen-2-yl)-1,2-oxazole
Synonyms: 5-(5-bromothiophen-2-yl)-1,2-oxazole
Molecular Weight (g/mol): 230.08
Molecular Formula: C7H4BrNOS
InChi Key: FCUZGSQWFAXBSB-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)