missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl 98.0+%, TCI America™
$158.44 - $633.78
Chemical Identifiers
| CAS | 142946-80-3 |
|---|---|
| Molecular Formula | C12H6F6N2 |
| Molecular Weight (g/mol) | 292.18 |
| MDL Number | MFCD24449717 |
| InChI Key | ZHMXQYAUGQASQM-UHFFFAOYSA-N |
| Synonym | 5,5′-Bis(trifluoromethyl)-2,2′-bipyridine |
| PubChem CID | 589903 |
| IUPAC Name | 5,5'-bis(trifluoromethyl)-2,2'-bipyridine |
| SMILES | FC(F)(F)C1=CN=C(C=C1)C1=NC=C(C=C1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B5449200MG
|
TCI America
B5449200MG |
200 mg |
Each for $158.44
|
|
|||||
|
B54491G
|
TCI America
B54491G |
1 g |
Each for $633.78
|
|
|||||
Chemical Identifiers
| 142946-80-3 | |
| 292.18 | |
| ZHMXQYAUGQASQM-UHFFFAOYSA-N | |
| 589903 | |
| FC(F)(F)C1=CN=C(C=C1)C1=NC=C(C=C1)C(F)(F)F |
| C12H6F6N2 | |
| MFCD24449717 | |
| 5,5′-Bis(trifluoromethyl)-2,2′-bipyridine | |
| 5,5'-bis(trifluoromethyl)-2,2'-bipyridine |
Specifications
| 142946-80-3 | |
| White-Yellow | |
| MFCD24449717 | |
| 5,5′-Bis(trifluoromethyl)-2,2′-bipyridine | |
| FC(F)(F)C1=CN=C(C=C1)C1=NC=C(C=C1)C(F)(F)F | |
| 292.18 | |
| 292.18 | |
| Crystalline Powder |
| 131°C | |
| C12H6F6N2 | |
| 200 mg | |
| ZHMXQYAUGQASQM-UHFFFAOYSA-N | |
| 5,5'-bis(trifluoromethyl)-2,2'-bipyridine | |
| 589903 | |
| ≥98.0% (T) | |
| 5,5′-Bis(trifluoromethyl)-2,2′-bipyridyl |
Safety and Handling
TSCA : No