accessibility menu, dialog, popup

UserName

CAS RN 142946-80-3

NFFFNFFF

CAS RN 142946-80-3

IUPAC Name: 5,5'-bis(trifluoromethyl)-2,2'-bipyridine
Synonyms: 5,5'-bis(trifluoromethyl)-2,2'-bipyridine
Molecular Weight (g/mol): 292.18
Molecular Formula: C12H6F6N2
InChi Key: ZHMXQYAUGQASQM-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)C1=NC=C(C=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results