missing translation for 'onlineSavingsMsg'
Learn More

5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane 98.0+%, TCI America™

Catalog No. b26825g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26825G Supplier TCI America Supplier No. B26825G

Chemical Identifiers

CAS 2425-41-4
Molecular Formula C12H16O4
Molecular Weight (g/mol) 224.256
InChI Key DHWCGYXHBIWIPM-UHFFFAOYSA-N
Synonym O,O′C-Benzylidenepentaerythritol, Monobenzalpentaerythritol
PubChem CID 17042
IUPAC Name [5-(hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl]methanol
SMILES C1C(COC(O1)C2=CC=CC=C2)(CO)CO

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Bis(hydroxymethyl)-2-phenyl-1,3-dioxane
RTECSNumber JH0300000
TSCA No