accessibility menu, dialog, popup

UserName

OHHOOO

CAS RN 2425-41-4

IUPAC Name: [5-(hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl]methanol
Synonyms: [5-(hydroxymethyl)-2-phenyl-1,3-dioxan-5-yl]methanol
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: DHWCGYXHBIWIPM-UHFFFAOYSA-N
SMILES: OCC1(CO)COC(OC1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results