missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5',6,6'-Tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindane 96.0+%, TCI America™
Supplier: TCI America T173725G
Specifications
| 5,5′,6,6′-Tetrahydroxy-3,3,3′,3′-tetramethyl-1,1′-spirobiindane | |
| White-Yellow | |
| MFCD00021235 | |
| 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, tts 5, 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindan, 1,1'-spirobi 1h-indene-5,5',6,6'-tetrol, 2,2',3,3'-tetrahydro-3,3,3',3'-tetramethyl, 1,1'-spirobi indane-5,6-diol , 3,3,3',3'-tetramethyl, 1,1,1',1'-tetramethyl-3,3'-spirobi 2h-indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2h,2'h-1,1'-spirobi indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2,2',3,3'-tetrahydro-1,1'-spirobi indene-5,5',6,6'-tetrol, 1,6-diol , 3,3,3',3'-tetramethyl | |
| CC1(CC2(CC(C3=CC(=C(C=C32)O)O)(C)C)C4=CC(=C(C=C41)O)O)C | |
| 340.419 | |
| 340.42 | |
| Crystalline Powder |
| 77-08-7 | |
| C21H24O4 | |
| 25 g | |
| POFMQEVZKZVAPQ-UHFFFAOYSA-N | |
| 1,1,1',1'-tetramethyl-3,3'-spirobi[2H-indene]-5,5',6,6'-tetrol | |
| 66162 | |
| ≥96.0% (HPLC,T) |
Chemical Identifiers
| 77-08-7 | |
| 340.419 | |
| POFMQEVZKZVAPQ-UHFFFAOYSA-N | |
| 66162 | |
| CC1(CC2(CC(C3=CC(=C(C=C32)O)O)(C)C)C4=CC(=C(C=C41)O)O)C |
| C21H24O4 | |
| MFCD00021235 | |
| 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobisindane, tts 5, 5,5',6,6'-tetrahydroxy-3,3,3',3'-tetramethyl-1,1'-spirobiindan, 1,1'-spirobi 1h-indene-5,5',6,6'-tetrol, 2,2',3,3'-tetrahydro-3,3,3',3'-tetramethyl, 1,1'-spirobi indane-5,6-diol , 3,3,3',3'-tetramethyl, 1,1,1',1'-tetramethyl-3,3'-spirobi 2h-indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2h,2'h-1,1'-spirobi indene-5,5',6,6'-tetrol, 3,3,3',3'-tetramethyl-2,2',3,3'-tetrahydro-1,1'-spirobi indene-5,5',6,6'-tetrol, 1,6-diol , 3,3,3',3'-tetramethyl | |
| 1,1,1',1'-tetramethyl-3,3'-spirobi[2H-indene]-5,5',6,6'-tetrol |
Safety and Handling
EINECSNumber : (4)-0587
TSCA : Yes