missing translation for 'onlineSavingsMsg'
Learn More

5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole, 95%

Catalog No. AAH5067703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5067703 1 g
AAH5067706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5067703 Supplier Thermo Scientific Chemicals Supplier No. H5067703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2251-79-8 | C8H5F3N4 | 214.151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2251-79-8
Molecular Formula C8H5F3N4
Molecular Weight (g/mol) 214.151
MDL Number MFCD00052525
InChI Key CCVCHQBLMDMSNN-UHFFFAOYSA-N
Synonym 5-4-trifluoromethyl phenyl-1h-tetrazole, 5-4-trifluoromethyl phenyl-2h-tetrazole, 5-4-trifluoromethyl phenyl-2h-1,2,3,4-tetrazole, 2h-tetrazole,5-4-trifluoromethyl phenyl, 5-4-trifluoromethyl phenyl-1h-1,2,3,4-tetrazole, 5-4-trifluoromethyl phenyl-2h-1,2,3,4-tetraazole, maybridge1_007473, acmc-1cqc2
PubChem CID 342565
IUPAC Name 5-[4-(trifluoromethyl)phenyl]-2H-tetrazole
SMILES C1=CC(=CC=C1C2=NNN=N2)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 214.15
Percent Purity 95%
Chemical Name or Material 5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only