missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde, 99%

Catalog No. AAH5048303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5048303 1 g
AAH5048306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5048303 Supplier Thermo Scientific Chemicals Supplier No. H5048303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38401-67-1 | C13H12O2S | 232.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38401-67-1
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
MDL Number MFCD00663639
InChI Key ODLOZSNXTQAWGQ-UHFFFAOYSA-N
Synonym 5-4-methoxyphenyl thiophene-2-carbaldehyde, 5-4-methoxyphenyl thiophene-2-carboxaldehyde, 5-4-methoxyphenyl-2-thiophenecarbaldehyde, acmc-1ahp9, rarechem ak ma k025, methoxyphenylthiophenecarbaldehyde, 2-formyl-5-4-methoxyphenyl thiophene, 5-4-methoxylphenyl thiophene-2-carbaldehyde, 5-4-methoxyphenyl-2-thiophenecarboxaldehyde, 5-4-methoxy-phenyl-thiophene-2-carbaldehyde
PubChem CID 2797800
IUPAC Name 5-(4-methoxyphenyl)thiophene-2-carbaldehyde
SMILES COC1=CC=C(C=C1)C1=CC=C(S1)C=O

Specifications

Quantity 1 g
Formula Weight 218.28
Percent Purity 99%
Chemical Name or Material 5-(4-Methoxyphenyl)thiophene-2-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only