accessibility menu, dialog, popup

UserName

CAS RN 38401-67-1

IUPAC Name: 5-(4-methoxyphenyl)thiophene-2-carbaldehyde
Synonyms: 5-(4-methoxyphenyl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 218.27
Molecular Formula: C12H10O2S
InChi Key: ODLOZSNXTQAWGQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(S1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results