missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxyphenyl)hydantoin 98.0+%, TCI America™

Catalog No. H08065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08065G 5 g
H080625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08065G Supplier TCI America Supplier No. H08065G
Only null left

Chemical Identifiers

CAS 2420-17-9
Molecular Formula C9H8N2O3
Molecular Weight (g/mol) 192.17
MDL Number MFCD00044002
InChI Key UMTNMIARZPDSDI-UHFFFAOYNA-N
PubChem CID 94233
IUPAC Name 5-(4-hydroxyphenyl)imidazolidine-2,4-dione
SMILES OC1=CC=C(C=C1)C1NC(=O)NC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 192.17
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Hydroxyphenyl)hydantoin
EINECSNumber (5)-5364
TSCA No