missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Chlorobutyl)hydantoin 97.0+%, TCI America™
$157.87 - $423.54
Chemical Identifiers
| CAS | 40126-55-4 |
|---|---|
| Molecular Formula | C7H11ClN2O2 |
| Molecular Weight (g/mol) | 190.627 |
| MDL Number | MFCD00059722 |
| InChI Key | ZKHLVOULDIXSCC-UHFFFAOYSA-N |
| PubChem CID | 229807 |
| IUPAC Name | 5-(4-chlorobutyl)imidazolidine-2,4-dione |
| SMILES | C(CCCl)CC1C(=O)NC(=O)N1 |
Chemical Identifiers
| 40126-55-4 | |
| 190.627 | |
| ZKHLVOULDIXSCC-UHFFFAOYSA-N | |
| 5-(4-chlorobutyl)imidazolidine-2,4-dione |
| C7H11ClN2O2 | |
| MFCD00059722 | |
| 229807 | |
| C(CCCl)CC1C(=O)NC(=O)N1 |
Specifications
| 40126-55-4 | |
| White-Yellow | |
| MFCD00059722 | |
| ZKHLVOULDIXSCC-UHFFFAOYSA-N | |
| 5-(4-chlorobutyl)imidazolidine-2,4-dione | |
| 229807 | |
| ≥97.0% (T) | |
| 5-(4-Chlorobutyl)hydantoin |
| 131°C | |
| C7H11ClN2O2 | |
| 5 g | |
| C(CCCl)CC1C(=O)NC(=O)N1 | |
| 190.627 | |
| 190.63 | |
| Crystalline Powder |
Safety and Handling
TSCA : No