accessibility menu, dialog, popup

UserName

CAS RN 40126-55-4

ClNHONHO

CAS RN 40126-55-4

IUPAC Name: 5-(4-chlorobutyl)imidazolidine-2,4-dione
Synonyms: 5-(4-chlorobutyl)imidazolidine-2,4-dione
Molecular Weight (g/mol): 190.63
Molecular Formula: C7H11ClN2O2
InChi Key: ZKHLVOULDIXSCC-UHFFFAOYNA-N
SMILES: ClCCCCC1NC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results