missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Bromophenyl)-4,6-dichloropyrimidine 98.0+%, TCI America™

Catalog No. B49501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B49501G Supplier TCI America Supplier No. B49501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146533-41-7
Molecular Formula C10H5BrCl2N2
Molecular Weight (g/mol) 303.97
MDL Number MFCD13151897
InChI Key WEEFLZORZXLIJE-UHFFFAOYSA-N
PubChem CID 19735262
IUPAC Name 5-(4-bromophenyl)-4,6-dichloropyrimidine
SMILES ClC1=NC=NC(Cl)=C1C1=CC=C(Br)C=C1

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 303.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Bromophenyl)-4,6-dichloropyrimidine