accessibility menu, dialog, popup

UserName

CAS RN 146533-41-7

NNClClBr

CAS RN 146533-41-7

IUPAC Name: 5-(4-bromophenyl)-4,6-dichloropyrimidine
Synonyms: 5-(4-bromophenyl)-4,6-dichloropyrimidine
Molecular Weight (g/mol): 303.97
Molecular Formula: C10H5BrCl2N2
InChi Key: WEEFLZORZXLIJE-UHFFFAOYSA-N
SMILES: ClC1=NC=NC(Cl)=C1C1=CC=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results