missing translation for 'onlineSavingsMsg'
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. t34851g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34851G 1 g
T34855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34851G Supplier TCI America Supplier No. T34851G
Only null left

Chemical Identifiers

CAS 1040281-83-1
Molecular Formula C11H15BO3S
Molecular Weight (g/mol) 238.108
MDL Number MFCD09266186
InChI Key OVGYVJMTHQRCDB-UHFFFAOYSA-N
Synonym 2-(5-Formyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Formyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 57415686
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C=O

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 1 g
Formula Weight 238.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde