accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CSO

CAS RN 1040281-83-1

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 238.11
Molecular Formula: C11H15BO3S
InChi Key: OVGYVJMTHQRCDB-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(S1)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results