missing translation for 'onlineSavingsMsg'
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7137768
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T34851G 1 g
T34855G 5 g
2 options

Catalog No. T34851G

Supplier: TCI America T34851G

Only null left

Chemical Identifiers

CAS 1040281-83-1
Molecular Formula C11H15BO3S
Molecular Weight (g/mol) 238.108
MDL Number MFCD09266186
InChI Key OVGYVJMTHQRCDB-UHFFFAOYSA-N
Synonym 2-(5-Formyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-Formyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 57415686
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(S2)C=O

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 1 g
Formula Weight 238.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde