missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile 98.0+%, TCI America™
$85.75 - $246.05
Chemical Identifiers
| CAS | 402718-29-0 |
|---|---|
| Molecular Formula | C12H15BN2O2 |
| Molecular Weight (g/mol) | 230.07 |
| MDL Number | MFCD07780755 |
| InChI Key | BOIKCRIMIQAFQJ-UHFFFAOYSA-N |
| Synonym | 3-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, nicotinonitril-5-boronic acid, 5-cyanopyridine-3-boronic acid pinacol ester, nicotinonitrile-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile?, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine |
| PubChem CID | 16414216 |
| IUPAC Name | 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile |
| SMILES | CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3514200MG
|
TCI America
T3514200MG |
200 mg |
Each for $85.75
|
|
|||||
|
T35141G
|
TCI America
T35141G |
1 g |
Each for $246.05
|
|
|||||
Chemical Identifiers
| 402718-29-0 | |
| 230.07 | |
| BOIKCRIMIQAFQJ-UHFFFAOYSA-N | |
| 16414216 | |
| CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)C#N |
| C12H15BN2O2 | |
| MFCD07780755 | |
| 3-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, nicotinonitril-5-boronic acid, 5-cyanopyridine-3-boronic acid pinacol ester, nicotinonitrile-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile?, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine | |
| 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile |
Specifications
| 402718-29-0 | |
| White | |
| MFCD07780755 | |
| 3-cyanopyridine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, nicotinonitril-5-boronic acid, 5-cyanopyridine-3-boronic acid pinacol ester, nicotinonitrile-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carbonitrile, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinonitrile?, 3-cyano-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine | |
| CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)C#N | |
| 230.07 | |
| 230.07 | |
| Crystalline Powder |
| 134°C | |
| C12H15BN2O2 | |
| 200 mg | |
| BOIKCRIMIQAFQJ-UHFFFAOYSA-N | |
| 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile | |
| 16414216 | |
| ≥98.0% (GC,T) | |
| 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile |
Safety and Handling
TSCA : No