missing translation for 'onlineSavingsMsg'
Learn More

5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride, 98%

Catalog No. AAJ6559622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6559622 100 g
AAJ6559614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6559622 Supplier Thermo Scientific Chemicals Supplier No. J6559622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A tricyclic serotnonin reuptake inhibitor | CAS: 549-18-8 | C20H24ClN | 313.869 g/mol

A tricyclic serotnonin reuptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 549-18-8
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.869
MDL Number MFCD00012537
InChI Key KFYRPLNVJVHZGT-UHFFFAOYSA-N
Synonym amitriptyline hydrochloride, amitriptyline hcl, annoyltin, tryptizol, domical, elavil hydrochloride, lentizol, saroten, amavil, ami-anelun
PubChem CID 11065
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b
SMILES CN(C)CCC=C1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 196°C to 197°C
Color White
Quantity 100 g
UN Number UN2811
Merck Index 14,487
Formula Weight 313.86
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-[3-(Dimethylamino)propylidene]dibenzosuberane hydrochloride
Hazard Category H301-H361d-H370
Hazard Statement GHS H Statement
H301-H311-H331-H334-H361-H315-H319-H317-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P310-P307+P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-964-6
RTECSNumber HO9450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only