missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Chlorophenyl)-1H-tetrazole, 98+%

Catalog No. AAL1069903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1069903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1069903 Supplier Thermo Scientific Chemicals Supplier No. L1069903
Only null left

CAS: 41421-28-7 | C7H4ClN4 | 179.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41421-28-7
Molecular Formula C7H4ClN4
Molecular Weight (g/mol) 179.59
MDL Number MFCD00040951
InChI Key WKXXXGMIBYWPGE-UHFFFAOYSA-N
Synonym 5-3-chlorophenyl-1h-tetrazole, 5-3-chlorophenyl-2h-tetrazole, 5-3-chloro-phenyl-2h-tetrazole, 5-3-chlorophenyl-1h-1,2,3,4-tetrazole, 5-3-chlorophenyl-2h-1,2,3,4-tetrazole, 5-3-chlorophenyl-1h-1,2,3,4-tetraazole, 5-3-chlorophenyl tetraazole, 5-chlorophenyltetrazole, acmc-20am9f, 5-3-chlorophenyl tetrazole
PubChem CID 592520
IUPAC Name 5-(3-chlorophenyl)-2H-tetrazole
SMILES ClC1=CC=CC(=C1)C1=N[N-]N=N1

Specifications

Melting Point 139°C to 140°C
Quantity 1 g
Beilstein 140081
Formula Weight 180.6
Percent Purity ≥98%
Chemical Name or Material 5-(3-Chlorophenyl)-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only