accessibility menu, dialog, popup

UserName

ClNNNN

CAS RN 41421-28-7

IUPAC Name: 5-(3-chlorophenyl)-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-(3-chlorophenyl)-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 179.59
Molecular Formula: C7H4ClN4
InChi Key: WKXXXGMIBYWPGE-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=N[N-]N=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results