missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Bromophenyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Chemical Identifiers

CAS 425379-69-7
Molecular Formula C10H9BrN2
Molecular Weight (g/mol) 237.1
MDL Number MFCD09065005
InChI Key RIYUQLBSAMXENJ-UHFFFAOYSA-N
Synonym 5-3-bromophenyl-1-methyl-1h-pyrazole, 5-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,5-3-bromophenyl-1-methyl
PubChem CID 22329560
IUPAC Name 5-(3-bromophenyl)-1-methylpyrazole
SMILES CN1C(=CC=N1)C2=CC(=CC=C2)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC60510CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60510DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60510DE
View Documents Promotion Details
N/A 5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

425379-69-7
237.1
RIYUQLBSAMXENJ-UHFFFAOYSA-N
22329560
CN1C(=CC=N1)C2=CC(=CC=C2)Br
C10H9BrN2
MFCD09065005
5-3-bromophenyl-1-methyl-1h-pyrazole, 5-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,5-3-bromophenyl-1-methyl
5-(3-bromophenyl)-1-methylpyrazole

Specifications

425379-69-7
100.0
C10H9BrN2
5-3-bromophenyl-1-methyl-1h-pyrazole, 5-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,5-3-bromophenyl-1-methyl
CN1C(=CC=N1)C2=CC(=CC=C2)Br
237.1
237.1
5-(3-Bromophenyl)-1-methyl-1H-pyrazole
96.5
97%
MFCD09065005
RIYUQLBSAMXENJ-UHFFFAOYSA-N
5-(3-bromophenyl)-1-methylpyrazole
22329560
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents