missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Bromophenyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC60510DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC60510DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

425379-69-7
237.1
RIYUQLBSAMXENJ-UHFFFAOYSA-N
22329560
CN1C(=CC=N1)C2=CC(=CC=C2)Br
C10H9BrN2
MFCD09065005
5-3-bromophenyl-1-methyl-1h-pyrazole, 5-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,5-3-bromophenyl-1-methyl
5-(3-bromophenyl)-1-methylpyrazole

Specifications

5-(3-Bromophenyl)-1-methyl-1H-pyrazole
96.5
97%
C10H9BrN2
MFCD09065005
RIYUQLBSAMXENJ-UHFFFAOYSA-N
5-(3-bromophenyl)-1-methylpyrazole
22329560
97%
425379-69-7
100.0
Amber Glass Bottle
5g
5-3-bromophenyl-1-methyl-1h-pyrazole, 5-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,5-3-bromophenyl-1-methyl
CN1C(=CC=N1)C2=CC(=CC=C2)Br
237.1
237.1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Promotions

Promotions