missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Furyl)pyridine-2-carbaldehyde, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC55104DA

 View more versions of this product

Catalog No. CC55104DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

5-(2-Furyl)pyridine-2-carbaldehyde
94.5
95%
C10H7NO2
1g
GUIKMKZMLLANPU-UHFFFAOYSA-N
5-(furan-2-yl)pyridine-2-carbaldehyde
42555941
95%
886851-45-2
100.0
Amber Glass Bottle
MFCD10700053
5-2-furyl pyridine-2-carbaldehyde, 5-furan-2-yl pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde,5-2-furanyl
C1=COC(=C1)C2=CN=C(C=C2)C=O
173.171
173.17

Chemical Identifiers

886851-45-2
173.171
GUIKMKZMLLANPU-UHFFFAOYSA-N
42555941
C1=COC(=C1)C2=CN=C(C=C2)C=O
C10H7NO2
MFCD10700053
5-2-furyl pyridine-2-carbaldehyde, 5-furan-2-yl pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde,5-2-furanyl
5-(furan-2-yl)pyridine-2-carbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions