missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%

Catalog No. AC446580050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC446580050 5 g
AC446580250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446580050 Supplier Thermo Scientific Chemicals Supplier No. 446580050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6299-85-0 | C6H4Cl2N2O2 | 207.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6299-85-0
Molecular Formula C6H4Cl2N2O2
Molecular Weight (g/mol) 207.02
InChI Key DQNQNLWKAGZNIT-UHFFFAOYSA-N
Synonym methyl 2,4-dichloropyrimidine-6-carboxylate, methyl 2,4-dichloro-pyrimidine-6-carboxylate, 2,6-dichloropyrimidine-4-carboxylic acid methyl ester, 4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester, 2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester, t6n cnj bg dvo1 fg wln, zlchem 33, pubchem23452, methyl2,6-dichloropyrimidine-4-carboxylate, 2,4-dichloro-6-methoxycarbonyl pyrimidine
PubChem CID 239689
IUPAC Name methyl 2,6-dichloropyrimidine-4-carboxylate
SMILES COC(=O)C1=CC(=NC(=N1)Cl)Cl

Specifications

Melting Point 55°C to 56°C
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material Methyl 2, 4-dichloropyrimidine-6-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and ke

RUO – Research Use Only