missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde 98.0+%, TCI America™
$182.68 - $549.89
Chemical Identifiers
| CAS | 881674-56-2 |
|---|---|
| Molecular Formula | C11H8FNO |
| Molecular Weight (g/mol) | 189.19 |
| MDL Number | MFCD11875867 |
| InChI Key | MQULPEUCGKEHEG-UHFFFAOYSA-N |
| PubChem CID | 46908593 |
| IUPAC Name | 5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde |
| SMILES | FC1=CC=CC=C1C1=CC(C=O)=CN1 |
Chemical Identifiers
| 881674-56-2 | |
| 189.19 | |
| MQULPEUCGKEHEG-UHFFFAOYSA-N | |
| 5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde |
| C11H8FNO | |
| MFCD11875867 | |
| 46908593 | |
| FC1=CC=CC=C1C1=CC(C=O)=CN1 |
Specifications
| 881674-56-2 | |
| Yellow | |
| MFCD11875867 | |
| MQULPEUCGKEHEG-UHFFFAOYSA-N | |
| 5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde | |
| 46908593 | |
| ≥98.0% (GC) | |
| 5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde |
| 133°C | |
| C11H8FNO | |
| 1 g | |
| FC1=CC=CC=C1C1=CC(C=O)=CN1 | |
| 189.19 | |
| 189.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No