accessibility menu, dialog, popup

UserName

HNOF

CAS RN 881674-56-2

IUPAC Name: 5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde
Synonyms: 5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde
Molecular Weight (g/mol): 189.19
Molecular Formula: C11H8FNO
InChi Key: MQULPEUCGKEHEG-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C1=CC(C=O)=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results