missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. F10745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10745G 5 g
F10741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10745G Supplier TCI America Supplier No. F10745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 881674-56-2
Molecular Formula C11H8FNO
Molecular Weight (g/mol) 189.19
MDL Number MFCD11875867
InChI Key MQULPEUCGKEHEG-UHFFFAOYSA-N
PubChem CID 46908593
IUPAC Name 5-(2-fluorophenyl)-1H-pyrrole-3-carbaldehyde
SMILES FC1=CC=CC=C1C1=CC(C=O)=CN1

Specifications

Melting Point 133°C
Color Yellow
Quantity 5 g
Formula Weight 189.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Fluorophenyl)pyrrole-3-carboxaldehyde