missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Bromophenyl)-1H-tetrazole 98.0+%, TCI America™

Catalog No. p-7119693
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B25775G 5 g
B257725G 25 g
2 options

Catalog No. B25775G

Supplier: TCI America B25775G

Only null left

Chemical Identifiers

CAS 73096-42-1
Molecular Formula C7H5BrN4
Molecular Weight (g/mol) 225.049
MDL Number MFCD00151799
InChI Key YHVBXKTXLJTDRI-UHFFFAOYSA-N
PubChem CID 560689
IUPAC Name 5-(2-bromophenyl)-2H-tetrazole
SMILES C1=CC=C(C(=C1)C2=NNN=N2)Br

Specifications

Melting Point 182°C
Color White-Yellow
Quantity 5 g
Synonym 5-2-bromophenyl-1h-tetrazole, 5-2-bromophenyl tetrazole, 5-2-bromo-phenyl-2h-tetrazole, 5-2-bromophenyl-2h-tetrazole, 5-2-bromophenyl-1h-1,2,3,4-tetrazole, 5-2-bromophenyl-2h-1,2,3,4-tetrazole, 5-2-bromophenyl-1h-tetraazole, 5-2-bromophenyl-1 h-tetrazole, 1h-tetrazole, 5-2-bromophenyl
Formula Weight 225.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(2-Bromophenyl)-1H-tetrazole