missing translation for 'onlineSavingsMsg'
Learn More

5,11-Dihydroindolo[3,2-b]carbazole 98.0+%, TCI America™

Catalog No. p-7128661
Change view
Click to view available options
Quantity:
1 g
200 mg
Catalog No. Quantity
D5124200MG 200 mg
D51241G 1 g
2 options

Catalog No. D5124200MG

Supplier: TCI America D5124200MG

Only null left

Chemical Identifiers

CAS 6336-32-9
Molecular Formula C18H12N2
Molecular Weight (g/mol) 256.31
MDL Number MFCD09879258
InChI Key YCPBCVTUBBBNJJ-UHFFFAOYSA-N
PubChem CID 95838
IUPAC Name 5H,11H-indolo[3,2-b]carbazole
SMILES N1C2=CC=CC=C2C2=CC3=C(C=C12)C1=C(N3)C=CC=C1

Specifications

Melting Point 460°C
Quantity 200 mg
Formula Weight 256.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,11-Dihydroindolo[3,2-b]carbazole