accessibility menu, dialog, popup

UserName

NHHN

CAS RN 6336-32-9

IUPAC Name: 5H,11H-indolo[3,2-b]carbazole
Synonyms: 5H,11H-indolo[3,2-b]carbazole
Molecular Weight (g/mol): 256.31
Molecular Formula: C18H12N2
InChi Key: YCPBCVTUBBBNJJ-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C2=CC3=C(C=C12)C1=C(N3)C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results