missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)porphyrin 95.0+%, TCI America™

Catalog No. t1495100mg
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
T1495100MG 100 mg
1 options

Catalog No. T1495100MG

Supplier: TCI America T1495100MG

Only null left

Chemical Identifiers

CAS 127812-08-2
Molecular Formula C52H38N4O12
Molecular Weight (g/mol) 910.892
MDL Number MFCD00191499
InChI Key BKLQXJKMYSCSFW-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)-21H,23H-porphine
PubChem CID 25076078
IUPAC Name 2-[4-[10,15,20-tris[4-(carboxymethoxy)phenyl]-21,24-dihydroporphyrin-5-yl]phenoxy]acetic acid
SMILES C1=CC(=CC=C1C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OCC(=O)O)C8=CC=C(C=C8)OCC(=O)O)C9=CC=C(C=C9)OCC(=O)O)N3)OCC(=O)O

Specifications

Color Black-Green
Quantity 100 mg
Formula Weight 910.89
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(4-carboxymethyloxyphenyl)porphyrin