accessibility menu, dialog, popup

UserName

CAS RN 127812-08-2

IUPAC Name: 2-(4-{7,12,17-tris[4-(carboxymethoxy)phenyl]-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaen-2-yl}phenoxy)acetic acid
Synonyms: 4-{7,12,17-tris[4-(carboxymethoxy)phenyl]-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaen-2-yl}phenoxyacetic acid
Molecular Weight (g/mol): 910.89
Molecular Formula: C52H38N4O12
InChi Key: BKLQXJKMYSCSFW-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=C(OCC(O)=O)C=C1)C1=CC=C(OCC(O)=O)C=C1)C1=CC=C(OCC(O)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results