missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetra(4-pyridyl)porphyrin 96.0+%, TCI America™

Catalog No. p-7137120
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22221G 1 g
T22225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22221G Supplier TCI America Supplier No. T22221G

Chemical Identifiers

CAS 16834-13-2
Molecular Formula C40H26N8
Molecular Weight (g/mol) 618.70
MDL Number MFCD00009626
InChI Key WQCKXOJXOKSXQZ-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine
PubChem CID 86073
IUPAC Name 2,7,12,17-tetrakis(pyridin-4-yl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
SMILES N1C2=CC=C1C(=C1C=CC(=N1)C(=C1C=CC(=N1)C(=C1NC(C=C1)=C2C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1

Specifications

Color Black-Purple
Quantity 1 g
Formula Weight 618.70
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetra(4-pyridyl)porphyrin